Learn More
Medchemexpress LLC Thymine-d4-1 | 156054-85-2 | 99.5% | 130.14 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW010450S410MG
Encompass_Preferred
Thymine-d4-1 is a deuterium-labeled version of Thymine, one of the four nucleobases found in DNA. Thymine can be a target for the actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM. This product is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium have been incorporated into drug molecules for quantitation during drug development, with deuterium specifically noted for its potential to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66488