Learn More
Medchemexpress LLC Thymine-d4 | 200496-79-3 | 98% | 130.14 | 50 MG
SDP
Catalog No. 5000366126
Encompass_Preferred
Thymine-d4 is the deuterium labeled Thymine, one of the four nucleobases in the nucleic acid of DNA. It can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-66126 Supplier Medchemexpress LLC Supplier No. 50MGHYW010450S
Thymine-d4 is the deuterium labeled Thymine, one of the four nucleobases in the nucleic acid of DNA. It can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs