Learn More
New England Biolabs, Inc. Thymine-DNA Glycosylase (TDG)-100 units
SDP
Catalog No. 5000332617
Encompass
Thymine-DNA glycosylase (TDG) catalyzes the hydrolysis of the N-glycosidic bond of thymine or uracil mispairs opposite guanine that arise from spontaneous deamination events of 5-methylcytosine (5mC) and cytosine, respectively.The excision of the mispaired thymine or uracil base leaves an abasic (AP) site that can then be chemically or enzymatically cleaved.
  • Active on dsDNA
  • Monofunctional glycosylase that removes dT or dU nucleobases mispaired with dG to create an AP site
  • Removes 5-formylcytosine, 5-carboxylcytosine, 5-hydroxymethyluracil, ethenocytosine, 5-fluorouracil
  • Isolated from a recombinant source
  • Supplied with 10X TDG Reaction Buffer
Catalog No. 50-003-32617 Supplier New England Biolabs, Inc. Supplier No. M0766S
Thymine-DNA glycosylase (TDG) catalyzes the hydrolysis of the N-glycosidic bond of thymine or uracil mispairs opposite guanine that arise from spontaneous deamination events of 5-methylcytosine (5mC) and cytosine, respectively.The excision of the mispaired thymine or uracil base leaves an abasic (AP) site that can then be chemically or enzymatically cleaved.
  • Active on dsDNA
  • Monofunctional glycosylase that removes dT or dU nucleobases mispaired with dG to create an AP site
  • Removes 5-formylcytosine, 5-carboxylcytosine, 5-hydroxymethyluracil, ethenocytosine, 5-fluorouracil
  • Isolated from a recombinant source
  • Supplied with 10X TDG Reaction Buffer