Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymine(Zidovudine Related Compound C) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015088 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5088 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5088 Supplier MilliporeSigma Supelco Supplier No. PHR1345250MG
Only null left
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 65-71-4
Physical Form Neat
Linear Formula C5H6N2O2
MDL Number MFCD00006026
Melting Point 320°C
Synonym Thymine; 2,4-Dihydroxy-5-methylpyrimidine; 5-Methyluracil
Type Certified Reference Material
Quantity 250 mg
Formula Weight 126.11
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.