Learn More
Cayman Chemical Thymohydroquinone, Purity: ≥98%, CAS: 2217-60-9, 50 mg
SDP
Catalog No. 502941478
Encompass_Preferred
A quinone with diverse biological activities; scavenges DPPH free radicals in a cell-free assay (IC50 = 2.4 μg/ml); has a Trolox equivalent value of 2.6 in an ORAC assay at 1.6-6.4 μg/ml; inhibits growth of A2780, OVCAR-8, and CIS-A2780 ovarian cancer cells (IC50s = 3.1, 8.9, and 9.8 μM, respectively) as well as immortalized human ovarian epithelial cells (IC50 = 14 μM); active against P. falciparum in vitro (IC50 = 15.9 μM)
Catalog No. 50-294-1478 Supplier Cayman Chemical Supplier No. 2755850 MG
A quinone with diverse biological activities; scavenges DPPH free radicals in a cell-free assay (IC50 = 2.4 μg/ml); has a Trolox equivalent value of 2.6 in an ORAC assay at 1.6-6.4 μg/ml; inhibits growth of A2780, OVCAR-8, and CIS-A2780 ovarian cancer cells (IC50s = 3.1, 8.9, and 9.8 μM, respectively) as well as immortalized human ovarian epithelial cells (IC50 = 14 μM); active against P. falciparum in vitro (IC50 = 15.9 μM)