Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018500 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8500 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8500 Supplier MilliporeSigma Supelco Supplier No. 72477500MGF
Add to Cart
Add to Cart

Thymol is a phenol compound and a major component of essential oil extracted from Thymus vulgaris L. medicinal plant. It exhibits antioxidant and antimicrobial properties and is widely used to treat bronchitis, gastritis, laryngitis, indigestion and dry coughs.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 89-83-8
Vapor Pressure 1 mmHg (64°C)
Density 0.965 g/mL (at 25°C)
Flash Point 101°C - Closed Cup
Physical Form Neat
Linear Formula 2-[(CH3)2 CH]C6H3-5-(CH3)OH
MDL Number MFCD00002309
Melting Point 48°C to 51°C; 49°C to 51°C
Boiling Point 232°C
Synonym 2-Isopropyl-5-methylphenol; 5-Methyl-2-(1-methylethyl)phenol; 5-Methyl-2-isopropylphenol; IPMP
Type Analytical Standard
Quantity 500 mg
Formula Weight 150.22
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.