Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol Blue Indicator, 0.4% w/v Aqueous, Reagents

Catalog No. 01333440
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-440 500 mL
01-333-441 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-440 Supplier Reagents Holdings Llc Supplier No. CT122000500A
Only null left
Add to Cart
Add to Cart

Thymol Blue Indicator, 0.4% w/v Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Thymol Blue Indicator, 0.4% w/v Aqueous
Boiling Point 100°C
CAS 7732-18-5, 62625-21-2
Synonym Thymolsulfonphthalein
Percent Purity 0.4%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.001
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.