Learn More
Specifications
Specifications
| CAS Max % | 100 |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Linear Formula | C27H29NaO5S |
| Decomposition Information | Sulfur compounds; Carbon monoxide; Carbon dioxide |
| Color | Gray/Green |
| Solubility Information | Soluble in water |
| Quantity | 5 g |
| Physical Form | Powder |
| Formula Weight | 488.58 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Product is not hazardous. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.