Learn More
Specifications
Specifications
| Chemical Name or Material | Thymol Blue |
| Name Note | Test Solution/Reagent Solution |
| CAS | 64-17-5, 67-56-1, 76-61-9 |
| CAS Min % | 94.0, 0.12, 4.0 |
| CAS Max % | 96.0, 0.13, 6.0 |
| Concentration | 0.1% (w/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Alcohol |
| Color | Dark Red |
| Quantity | 120 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
