Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019589 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9589 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9589 Supplier MilliporeSigma Supelco Supplier No. PHR11341G
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 89-83-8
Vapor Pressure 1 mmHg (64°C)
Density 0.965 g/mL (at 25°C)
Flash Point 101°C - Closed Cup
Physical Form Neat
Linear Formula 2-[(CH3)2 CH]C6H3-5-(CH3)OH
MDL Number MFCD00002309
Melting Point 48°C to 51°C
Boiling Point 232°C
Synonym 2-Isopropyl-5-methylphenol; 5-Methyl-2-(1-methylethyl)phenol; 5-Methyl-2-isopropylphenol; IPMP
Type Certified Reference Material
Quantity 1 g
Formula Weight 150.22
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.