Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein, 0.1% in Ethanol, Certified, LabChem™

Catalog No. LC260757 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC260757 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC260757 Supplier LabChem, Inc. Supplier No. LC260757
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Thymolphthalein
Name Note 0.1% in Ethanol
CAS Max % 90.4, 5, 4.5, 0.1
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
CAS 67-63-0, 67-56-1, 125-20-2
Color Colorless
Packaging Poly Bottle
Linear Formula C28H30O4
Quantity 125 mL
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Solubility Information Soluble in water
Formula Weight 430.5
Grade Certified
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.