Learn More
Abcam Thymolphthalein, Acid-base pH indicator (colorless-blue), 25G
Catalog No. 502704843
Encompass_Preferred
MW 430.5 g/mol. Acid-base pH indicator. The transition range is around pH 9.3–10.5. Below this pH, it is colorless; above, it is blue. Acidic form retains hydrogen on each hydroxyl group.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4843 Supplier Abcam Supplier No. AB14620725G
MW 430.5 g/mol. Acid-base pH indicator. The transition range is around pH 9.3–10.5. Below this pH, it is colorless; above, it is blue. Acidic form retains hydrogen on each hydroxyl group.
The product is subject to the following: Abcam Restricted Use Statement