Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein, ACS reagent, Indicator (pH 8.8-10.5), MilliporeSigma™ Supelco™

Catalog No. 111012842 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-101-2842 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2842 Supplier MilliporeSigma Supelco Supplier No. 8936010G

missing translation for 'validMainframeMsg'

Thymolphthalien is an acid-base indicator which is colourless in acidic form and deep blue in basic form. Acidic form retains hydrogen on each hydroxyl group. It is suitable in preparing disappearing ink.

TRUSTED_SUSTAINABILITY

Specifications

CAS 125-20-2
Melting Point 248°C to 254°C; 251°C to 253°C
Linear Formula C28H30O4
MDL Number MFCD00005909
Quantity 10 g
Synonym 5 ',5 ''-Diisopropyl-2 ',2 ''-dimethylphenolphthalein
Formula Weight 430.54
Grade ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.