Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein ACS reagent, indicator (pH 8.8-10.5), MilliporeSigma™ Supelco™

Catalog No. 111012843 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2843 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2843 Supplier MilliporeSigma Supelco Supplier No. 8936050G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymolphthalien is an acid-base indicator which is colourless in acidic form and deep blue in basic form. Acidic form retains hydrogen on each hydroxyl group. It is suitable in preparing disappearing ink.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-20-2
Molecular Weight (g/mol) 430.54
MDL Number MFCD00005909
Synonym 5 ′,5 ′′-Diisopropyl-2 ′,2 ′′-dimethylphenolphthalein

Specifications

Melting Point 248°C to 254°C; 251°C to 253°C
Linear Formula C28H30O4
Quantity 50 g
Formula Weight 430.54
Grade ACS Reagent
Chemical Name or Material Thymolphthalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.