Learn More
CPC Scientific H-Arg-Lys-Asp-Val-Tyr-OH 1MG
SDP
Supplier:  CPC Scientific THYM001A
Encompass
Thymopentin (TP5) is an active fragment of thymopoietin (TP) that reduces endocrine and behavioral responses to experimental stress. TP5 has immunomodulatory effects by suppression of proliferation of HL-60 cells.SEQUENCE: H-Arg-Lys-Asp-Val-Tyr-OHONE-LETTER SEQUENCE: RKDVYMOLECULAR FORMULA: C30H49N9O9MOLECULAR WEIGHT: 679.77STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [177966-81-3]SYNONYMS: Thymopoietin-5, Thymopoietin (32-36) (human, mouse), (Asp??)-Thymopoietin III (32-36) (bovine), TP-5, Thymopoietin I/II (32-36) (bovine)RESEARCH AREA: ImmunomodulationREFERENCES:G.Goldstein et al., Biol. Psychiatry, 48, 65 (2000)Y.-Z.Fan et al., Biochim. Biophys. Acta, 1763, 1059 (2006)SKU(s): THYM-001A, THYM-001B, THYM-001C
Catalog No. 50-211-0571