Learn More
Bachem Thymopentin Acetate
SDP
Supplier:  Bachem 4002968.0025
Encompass_Preferred
Thymopentin Acetate, H-Arg-Lys-Asp-Val-Tyr-OH, 177966-81-3, C30H49N9O9, Mr 679.77, 25mg. TP-5, Thymopoietin (32-36) (human, mouse), (Asp34)-Thymopoietin III (32-36) (bovine), Thymopoietin-5, Thymopoietin I/II (32-36) (bovine). Thymopentin (TP5), an active fragment of thymopoietin (TP), reduces endocrine and behavioral responses to experimental stress, possibly by lowering plasma TP (pTP) levels. The immunomodulatory peptide suppresses proliferation and induces differentiation in HL-60 cells.
Catalog No. 50-383-123