Learn More
Medchemexpress LLC Thymopentin | 69558-55-0 | 10 MG
SDP
Catalog No. 5000419014
Encompass_Preferred
Thymopentin is a biologically active peptide primarily secreted by the epithelial cells of the thymic cortex and medulla. It acts as an effective immunomodulatory agent.
  • Effective immunomodulatory agent.
  • Short plasma half-life of 30 seconds.
  • Enhances the generation of T-cell lineage.
  • Supports T-cell lineage derived from human embryonic stem cells.
Catalog No. 50-004-19014 Supplier Medchemexpress LLC Supplier No. HYN712210MG
Thymopentin is a biologically active peptide primarily secreted by the epithelial cells of the thymic cortex and medulla. It acts as an effective immunomodulatory agent.
  • Effective immunomodulatory agent.
  • Short plasma half-life of 30 seconds.
  • Enhances the generation of T-cell lineage.
  • Supports T-cell lineage derived from human embryonic stem cells.