Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thymopentin | 69558-55-0 | 5 MG
SDP

Catalog No. 5000418523
Encompass_Preferred

Thymopentin is a biologically active peptide that is primarily secreted by the epithelial cells of the thymic cortex and medulla. It functions as an effective immunomodulatory agent, characterized by a short plasma half-life of 30 seconds. This compound has been shown to enhance the generation of T-cell lineage derived from human embryonic stem cells (hESCs).

  • Purity: 99.94%
  • Molecular weight: 679.77
  • Formula: C30H49N9O9
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Arg-Lys-Asp-Val-Tyr (RKDVY)
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Endogenous metabolite

Catalog No. 50-004-18523 Supplier Medchemexpress LLC Supplier No. HYN71225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymopentin is a biologically active peptide that is primarily secreted by the epithelial cells of the thymic cortex and medulla. It functions as an effective immunomodulatory agent, characterized by a short plasma half-life of 30 seconds. This compound has been shown to enhance the generation of T-cell lineage derived from human embryonic stem cells (hESCs).

  • Purity: 99.94%
  • Molecular weight: 679.77
  • Formula: C30H49N9O9
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Arg-Lys-Asp-Val-Tyr (RKDVY)
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.