Learn More
Medchemexpress LLC HY-N7122A 5mg Medchemexpress, Thymopentin (acetate) CAS:89318-88-7 Purity:>98%
SDP
Catalog No. 501970506
Encompass_Preferred
Medchemexpress, HY-N7122A 5mg Thymopentin (acetate) CAS:89318-88-7 Thymopentin acetate is a biologically active peptide secreted mainly by the epithelial cells of thymic cortex and medulla. Thymopentin acetate is an effective immunomodulatory agent with a short plasma half-life of 30 seconds. Thymopentin acetate enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0506 Supplier Medchemexpress LLC Supplier No. HYN7122A5MG
Medchemexpress, HY-N7122A 5mg Thymopentin (acetate) CAS:89318-88-7 Thymopentin acetate is a biologically active peptide secreted mainly by the epithelial cells of thymic cortex and medulla. Thymopentin acetate is an effective immunomodulatory agent with a short plasma half-life of 30 seconds. Thymopentin acetate enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.