Learn More
Medchemexpress LLC Thymopentin (acetate) | 89318-88-7 | 10 MG
SDP
Catalog No. 5000418322
Encompass_Preferred
Thymopentin acetate is a biologically active peptide secreted mainly by the epithelial cells of the thymic cortex and medulla. It is an effective immunomodulatory agent with a short plasma half-life of 30 seconds. It enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs).
  • Biologically active peptide
  • Effective immunomodulatory agent
  • Short plasma half-life of 30 seconds
  • Enhances generation of T-cell lineage from human embryonic stem cells (hESCs)
  • High purity (99.95%)
Catalog No. 50-004-18322 Supplier Medchemexpress LLC Supplier No. HYN7122A10MG
Thymopentin acetate is a biologically active peptide secreted mainly by the epithelial cells of the thymic cortex and medulla. It is an effective immunomodulatory agent with a short plasma half-life of 30 seconds. It enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs).
  • Biologically active peptide
  • Effective immunomodulatory agent
  • Short plasma half-life of 30 seconds
  • Enhances generation of T-cell lineage from human embryonic stem cells (hESCs)
  • High purity (99.95%)