Learn More
Medchemexpress LLC Thymopentin acetate | 89318-88-7 | 50 MG
SDP
Catalog No. 5000417677
Encompass_Preferred
Thymopentin acetate is a biologically active peptide primarily secreted by the epithelial cells of the thymic cortex and medulla. It functions as an effective immunomodulatory agent, characterized by a short plasma half-life of 30 seconds. This compound also enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs).
  • Biologically active peptide
  • Functions as an effective immunomodulatory agent
  • Characterized by a short plasma half-life of 30 seconds
  • Enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs)
Catalog No. 50-004-17677 Supplier Medchemexpress LLC Supplier No. HYN7122A50MG
Add to Cart
Add to Cart
Thymopentin acetate is a biologically active peptide primarily secreted by the epithelial cells of the thymic cortex and medulla. It functions as an effective immunomodulatory agent, characterized by a short plasma half-life of 30 seconds. This compound also enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs).
  • Biologically active peptide
  • Functions as an effective immunomodulatory agent
  • Characterized by a short plasma half-life of 30 seconds
  • Enhances the generation of T-cell lineage derived from human embryonic stem cells (hESCs)