Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymoquinone, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014306 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4306 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4306 Supplier MilliporeSigma Supelco Supplier No. 03416100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymoquinone is employed in preclinical studies as neuroprotective and anticonvulsant agent.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 490-91-5
Flash Point 104°C
Linear Formula C10H12O2
MDL Number MFCD00001602
Melting Point 45°C to 47°C
Boiling Point 230°C to 232°C
Synonym 2-Isopropyl-5-methyl-1,4-benzoquinone
Type Analytical Standard
Quantity 100 mg
Formula Weight 164.20
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.