Learn More
Selleck Chemical LLC Thymosin β4 Acetate (0.95) 25mg 77591-33-4 Ac-Ser-Asp-Lys-Pro-Asp-Met-Ala-Glu-Ile-Glu-Lys-Phe-Asp-Lys-Ser-Lys-Leu-Lys-Lys-Thr-Glu-Thr-Gln-Glu-Lys-Asn-Pro-Leu-Pro-Ser-Lys-Glu-Thr-Ile-Glu-Gln-Glu-Lys-Gln-Ala-Gly-Glu-Ser-OH
SDP
Supplier:  Selleck Chemical LLC P103625MG
Encompass_Preferred
Thymosin β4 Acetate (0.95) is a 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide.
Catalog No. 50-194-8370