Learn More
CPC Scientific Ac-Ser-Asp-Ala-Ala-Val-Asp-Thr-Ser-Ser-Glu-Ile-Thr-Thr-Lys-Asp-Leu-Lys-Glu-Lys-Lys-Glu-Val-Val-Glu-Glu-Ala-Glu-Asn-OH 1MG
SDP
Supplier:  CPC Scientific THYM004B
Encompass
Thymosin -1 is 28-amino acid fragment of a 113-amino acid precursor peptide, prothymosin . The 28-mer is involved in restoring immune function in animals that lack thymus glands and has been found to enhance cell-mediated immunity in both animal and human models.SEQUENCE: Ac-Ser-Asp-Ala-Ala-Val-Asp-Thr-Ser-Ser-Glu-Ile-Thr-Thr-Lys-Asp-Leu-Lys-Glu-Lys-Lys-Glu-Val-Val-Glu-Glu-Ala-Glu-Asn-OHONE-LETTER SEQUENCE: Ac-SDAAVDTSSEITTKDLKEKKEVVEEAENMOLECULAR FORMULA: C129H215N33O55MOLECULAR WEIGHT: 3108.32STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [62304-98-7]SYNONYMS: Thymalfasin, Thymosin -1RESEARCH AREA: ImmunomodulationREFERENCES:M.B. Sztein et al, Springer Semin. Immunopathol., 9, 1(1986)SKU(s): THYM-004A, THYM-004B, THYM-004C
Catalog No. 50-211-0577