Learn More
CPC Scientific Ac-Ala-Asp-Lys-Pro-Asp-Met-Gly-Glu-Ile-Ala-Ser-Phe-Asp-Lys-Ala-Lys-Leu-Lys-Lys-Thr-Glu-Thr-Gln-Glu-Lys-Asn-Thr-Leu-Pro-Thr-Lys-Glu-Thr-Ile-Glu-Gln-Glu-Lys-Arg-Ser-Glu-Ile-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific THYM006B
Encompass
ONE-LETTER SEQUENCE: Ac-ADKPDMGEIASFDKAKLKKTETQEKNTLPTKETIEQEKRSEISMOLECULAR FORMULA: C211H353N57O76S1MOLECULAR WEIGHT:4936.53STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [88160-82-1], RESEARCH AREA: ImmunomodulationREFERENCES: V.McCreary et al., Biochem. Biophys. Res. Commun., 152, 862 (1988)
Catalog No. 50-211-0582