Learn More
Medchemexpress LLC Thymotrinan | 00-00-0 | MFCD00237286 | 98.7% | 531.48 g·mol⁻¹ | C18H32F3N7O8 | 5 MG
SDP
Catalog No. 5000240747
Encompass_Preferred
Thymotrinan TFA is the trifluoroacetate (TFA) salt of a biologically active fragment of the thymus hormone thymopoietin. It exhibits immunomodulatory activity similar to TP5, affecting both humoral and cellular immune responses, and is supplied as a white to off-white solid for research use. Manufacturer storage and handling guidelines should be followed.
Catalog No. 50-002-40747 Supplier Medchemexpress LLC Supplier No. HY106765A5MG
Thymotrinan TFA is the trifluoroacetate (TFA) salt of a biologically active fragment of the thymus hormone thymopoietin. It exhibits immunomodulatory activity similar to TP5, affecting both humoral and cellular immune responses, and is supplied as a white to off-white solid for research use. Manufacturer storage and handling guidelines should be followed.