Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thymotrinan trifluoroacetate | 00-00-0 | 98.7% | 531.48 g·mol⁻¹ | C18H32F3N7O8 | 25 MG
SDP

Catalog No. 5000242116
Encompass_Preferred

Thymotrinan TFA is a biologically active fragment of the thymus hormone thymopoietin, provided as the trifluoroacetate (TFA) salt for research use. It modulates immune responses, affecting both humoral and cellular immunity, and is supplied as a purified synthetic peptide for laboratory studies.

  • Immunomodulating peptide that affects humoral and cellular responses.
  • Supplied as the trifluoroacetate salt for improved stability and handling.
  • High purity suitable for in vitro and preclinical research.
  • Available in small-scale quantities for peptide research workflows.
  • Molecular weight 531.48 g·mol⁻¹; formula C18H32F3N7O8.

Catalog No. 50-002-42116 Supplier Medchemexpress LLC Supplier No. HY106765A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymotrinan TFA is a biologically active fragment of the thymus hormone thymopoietin, provided as the trifluoroacetate (TFA) salt for research use. It modulates immune responses, affecting both humoral and cellular immunity, and is supplied as a purified synthetic peptide for laboratory studies.

  • Immunomodulating peptide that affects humoral and cellular responses.
  • Supplied as the trifluoroacetate salt for improved stability and handling.
  • High purity suitable for in vitro and preclinical research.
  • Available in small-scale quantities for peptide research workflows.
  • Molecular weight 531.48 g·mol⁻¹; formula C18H32F3N7O8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.