Learn More
Cayman Chemical ThymulInacetate hydrate 10mg
SDP
Catalog No. 502946357
Encompass_Preferred
A peptide hormone; primarily produced by thymic epithelial cells; potentiates mitogen-induced proliferation of IL-2 receptor-positive mature T cells; has anti-inflammatory effects in various rodent models of inflammation; suppresses autoantibody production in a mouse model of reovirus-induced thyroiditis; prevents endotoxin-induced hyperalgesia in rats
Catalog No. 50-294-6357 Supplier Cayman Chemical Supplier No. 3341810 MG
A peptide hormone; primarily produced by thymic epithelial cells; potentiates mitogen-induced proliferation of IL-2 receptor-positive mature T cells; has anti-inflammatory effects in various rodent models of inflammation; suppresses autoantibody production in a mouse model of reovirus-induced thyroiditis; prevents endotoxin-induced hyperalgesia in rats