Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Thymyl acetate | 95% | CAS 528-79-0 | C12H16O2 | 1g

Catalog No. 5001008709
Encompass_Preferred

Thymyl acetate | 95% | CAS 528-79-0 | C12H16O2 | 1g

Catalog No. 50-010-08709 Supplier Aobchem Supplier No. A2749731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymyl acetate | 95% | CAS 528-79-0 | C12H16O2 | 1g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.