Learn More
Aviva Systems Biology Corp THYN1 Antibody - middle region (ARP54988_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54988_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THYN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human THYN1. Alternate names: HSPC144, MDS012, MGC12187, MY105, THY28, THY28KDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61387