Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7038MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IHC. Conjugate/Tag: Unconjugated. Clone: SPM221. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Thyroid. Cellular Staining: Cytoplasmic, secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human thyroid follicular cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 660kDa (Dimeric Form).Chromosome: 8q24.22. Gene Symbol: TG. Gene ID: 7038. Ensemble: ENSG00000042832. Uniprot: NP_003226.
Catalog No. 50-188-5204