Learn More
Biogenex Laboratories Thyroglobulin : 2H11 : 6mL
SDP
Supplier:  Biogenex Laboratories AM0325M
Encompass_Preferred
Thyroglobulin is a 19S glycoprotein with a molecular mass ofapproximately 650 kD. It constitutes 85-100% of the total of all thyroidiodoproteins. Immunohistochemical studies of thyroid carcinomas haverevealed that a high portion of differentiated thyroid carcinomassynthesize Thyroglobulin. Positive thyroglobulin staining indicatesthyroidal origin of the tumor. Immunohistochemical and electronmicroscopic findings have disclosed a wide range of cellulardifferentiation in thyroid adenomas.
Catalog No. 50-197-8810