Learn More
Diagnostic Biosystems THYROGLOBULIN Thyroglobulin (1D4)
Catalog No. 502407339
Encompass_Preferred
This antibody recognizes thyroglobulin in hyperplastic and neoplastic thyroid. Thyroglobulin is a protein of 670 kDa consisting of two identical subunits. Thyroglobulin is synthesized by the follicular epithelial cells of the thyroid.
Catalog No. 50-240-7339 Supplier Diagnostic Biosystems Supplier No. PDM576
This antibody recognizes thyroglobulin in hyperplastic and neoplastic thyroid. Thyroglobulin is a protein of 670 kDa consisting of two identical subunits. Thyroglobulin is synthesized by the follicular epithelial cells of the thyroid.