Learn More
Biocare Medical, LLC Thyroglobulin Cocktail Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM022AA
Encompass_Preferred
Thyroglobulin antibody cocktail has been shown to react with human thyroglobulin, staining thyroglobulin in follicular epithelial cells as well as colloid tissue. Clones 2H11 6E1 have been shown to be useful in positive identification of both papillary and follicular types of thyroid carcinomas. Demonstration of thyroglobulin staining via immunohistochemistry in a metastatic lesion establishes the thyroid origin of the tumor. Poorly differentiated carcinomas of the thyroid are frequently thyroglobulin negative. Adenocarcinomas from a non-thyroid origin are not reactive.
Catalog No. 50-830-08