Learn More
Cayman Chemical ThyroId Hormon Receptr nta 5mg
SDP
Catalog No. 502966633
Encompass_Preferred
An orally active selective inhibitor of Cdk4 (IC50 11 nM) and Cdk6 (IC50 16 nM) with antiproliferative activity against retinoblastoma-positive tumor cells blocking retinoblastoma phosphorylation and inducing G1 arrest used to demonstrate a role for insulin-activated cyclin D1-Cdk4 signaling in the control of glucose metabolism
Catalog No. 50-296-6633 Supplier Cayman Chemical Supplier No. 172485 MG
An orally active selective inhibitor of Cdk4 (IC50 11 nM) and Cdk6 (IC50 16 nM) with antiproliferative activity against retinoblastoma-positive tumor cells blocking retinoblastoma phosphorylation and inducing G1 arrest used to demonstrate a role for insulin-activated cyclin D1-Cdk4 signaling in the control of glucose metabolism