Learn More
Cayman Chemical ThyroId Hormon RECPTR nta 10mg
SDP
Catalog No. 502967449
Encompass_Preferred
A natural anthocyanin that scavenges superoxide anion radicals (IC50 69 UM) and protects neurons from oxidative stress inhibits the ADP-ribosyl cyclase CD38 (IC50 6.3 UM) preventing the metabolism of NAD and NADP
Catalog No. 50-296-7449 Supplier Cayman Chemical Supplier No. 1724810 MG
A natural anthocyanin that scavenges superoxide anion radicals (IC50 69 UM) and protects neurons from oxidative stress inhibits the ADP-ribosyl cyclase CD38 (IC50 6.3 UM) preventing the metabolism of NAD and NADP