Learn More
Bioss Thyroid Hormone Receptor 1/
SDP
Catalog No. 5000064739
Encompass_Preferred
Thyroid Hormone Receptor 1/ 2-Isotype Primary Antibody Unconjugated Host Species Mouse Target Species Human Rat Clonality Monoclonal Applications WB 1718Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol
Catalog No. 50-000-64739 Supplier Bioss Supplier No. BSM70286M
Thyroid Hormone Receptor 1/ 2-Isotype Primary Antibody Unconjugated Host Species Mouse Target Species Human Rat Clonality Monoclonal Applications WB 1718Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol