Learn More
Bioss Thyroid hormone receptor alpha
SDP
Catalog No. 5000066316
Encompass_Preferred
Thyroid hormone receptor alpha Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66316 Supplier Bioss Supplier No. BSM62198R
Thyroid hormone receptor alpha Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol