Learn More
Bioss Thyroid Hormone Receptor -Is
SDP
Supplier:  Bioss BSM70288M
Encompass_Preferred
Thyroid Hormone Receptor -Isotype Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB 2386Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol
Catalog No. 50-000-64740