Learn More
Medchemexpress LLC Thyroid peroxidase antibody | 10 UL
SDP
Catalog No. 5000441705
Encompass_Preferred
Thyroid Peroxidase Antibody (YA1480) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Thyroid Peroxidase. It is intended for research use only.
  • Targets thyroid peroxidase
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Human species reactivity
  • Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Applications: Western blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunoprecipitation (IP)
Catalog No. 50-004-41705 Supplier Medchemexpress LLC Supplier No. HYP8173510UL
Add to Cart
Add to Cart
Thyroid Peroxidase Antibody (YA1480) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Thyroid Peroxidase. It is intended for research use only.
  • Targets thyroid peroxidase
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Human species reactivity
  • Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Applications: Western blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunoprecipitation (IP)