Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thyroid peroxidase antibody (YA1480) | 50 UL
SDP

Catalog No. 5000440843
Encompass_Preferred

This rabbit-derived, non-conjugated IgG monoclonal antibody targets Thyroid Peroxidase and is intended for research use only. It is supplied as a liquid in a specialized buffer solution for stability.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Reacts with human species
  • Immunogen is a synthesized peptide derived from human TPO aa880-933
  • Endogenous sensitivity
  • Purity by affinity chromatography
  • Non-conjugated and unmodified
  • Isotype: IgG
  • Applications include WB (1:500-1:1000), IHC-P (1:50-1:100), and IP (1:40)
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Predicted and observed molecular weight: 103 kDa

Catalog No. 50-004-40843 Supplier Medchemexpress LLC Supplier No. HYP8173550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit-derived, non-conjugated IgG monoclonal antibody targets Thyroid Peroxidase and is intended for research use only. It is supplied as a liquid in a specialized buffer solution for stability.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Reacts with human species
  • Immunogen is a synthesized peptide derived from human TPO aa880-933
  • Endogenous sensitivity
  • Purity by affinity chromatography
  • Non-conjugated and unmodified
  • Isotype: IgG
  • Applications include WB (1:500-1:1000), IHC-P (1:50-1:100), and IP (1:40)
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Predicted and observed molecular weight: 103 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.