Learn More
MP Biomedical™ Thyroid Stimulating Hormone (TSH) RIA Kit for Rats
Shop All MP Biomedicals, Inc ProductsDescription
Specifications
Specifications
| Description | Intended for the measurement of prolactin in a rat sample. |
| Product Type | Kit and Assay |
| Sample Type | Blood, Cell Culture, Tissue |
| For Use With (Application) | Animal Research |
| Quantity | 125 Determinations |
| Regulatory Status | For research use only (RUO) |
| Sample Volume | 100 μL |
| Storage Requirements | 2°C to 8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.