Learn More
CPC Scientific Pyr-His-Pro-NH2 (trifluoroacetate salt) 50MG
SDP
Supplier:  CPC Scientific TRHP001A
Encompass
SEQUENCE: Pyr-His-Pro-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Pyr-HP-NH2MOLECULAR FORMULA: C16H22N6O4MOLECULAR WEIGHT: 362.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [24305-27-9]SYNONYMS: RESEARCH AREA: HormonalREFERENCES:Ohki, K. et al. Chem. Pharm. Bull. 45, 194 (1997)Laakkonen, L. et al. Mol. Pharmacol. 49, 1092 (1996)Scanes, C. et al. Acta Endocrinol. (Copenh) 97, 448 (1981).SKU(s): TRHP-001A
Catalog No. 50-211-0585