Learn More
Medchemexpress LLC Thyrotropin-releasing hormone (TRH), free acid | 24769-58-2 | 99.98% | 363.37 | 50 MG
SDP
Catalog No. 5000422178
Encompass_Preferred
Thyrotropin-releasing hormone (TRH), free acid is a highly conserved neuropeptide that plays a crucial role in the hormonal control of thyroid-stimulating hormone (TSH) levels and also exhibits neuromodulatory functions. It is an endogenous metabolite.
  • Highly conserved neuropeptide.
  • Exerts hormonal control of thyroid-stimulating hormone (TSH) levels.
  • Possesses neuromodulatory functions.
  • Agonist activity at mouse TRH-R1 and TRH-R2 receptors, assessed by Ca2+ release and IP1 formation.
  • Induces a temporary, dose-dependent rise in body temperature in rats.
  • Hyperthermic action may involve thyroid hormone release.
Catalog No. 50-004-22178 Supplier Medchemexpress LLC Supplier No. HYP152950MG
Thyrotropin-releasing hormone (TRH), free acid is a highly conserved neuropeptide that plays a crucial role in the hormonal control of thyroid-stimulating hormone (TSH) levels and also exhibits neuromodulatory functions. It is an endogenous metabolite.
  • Highly conserved neuropeptide.
  • Exerts hormonal control of thyroid-stimulating hormone (TSH) levels.
  • Possesses neuromodulatory functions.
  • Agonist activity at mouse TRH-R1 and TRH-R2 receptors, assessed by Ca2+ release and IP1 formation.
  • Induces a temporary, dose-dependent rise in body temperature in rats.
  • Hyperthermic action may involve thyroid hormone release.