Learn More
Cayman Chemical Thyrotropin-Releasing Hormone, Purity: ≥98%, CAS: 24305-27-9, 100 mg
SDP
Catalog No. 502993933
Encompass_Preferred
A tropic hormone that stimulates TSH and prolactin release; binds to the human and rat TRHR (IC50s = 25 and 198 nM, respectively); it dose-dependently increases TSH release from rat anterior pituitary slices and prolactin release from cultured anterior pituitary cells; increases TSH and prolactin plasma levels in vivo in rats (0.01 mg/kg)
Catalog No. 50-299-3933 Supplier Cayman Chemical Supplier No. 22917100 MG
A tropic hormone that stimulates TSH and prolactin release; binds to the human and rat TRHR (IC50s = 25 and 198 nM, respectively); it dose-dependently increases TSH release from rat anterior pituitary slices and prolactin release from cultured anterior pituitary cells; increases TSH and prolactin plasma levels in vivo in rats (0.01 mg/kg)