Learn More
MOLECULAR DEPOT LLC Thyroxine BSA Conjugate - 5 mg
Catalog No. 5000013473
Encompass_Preferred
Thyroxine BSA Conjugate (5 mg) is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Thyroxine (also known L-Thyroxine Levothyroxine T4 and Tetraiodothyronine ) A carbon spacer arm of 10 0 A links BSA to the Thyroxine moiety Each BSA molecule contains several Thyroxine units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline pH 7 4 For in vitro research use only Not for human or animal use
Catalog No. 50-000-13473 Supplier MOLECULAR DEPOT LLC Supplier No. P2010027
Thyroxine BSA Conjugate (5 mg) is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Thyroxine (also known L-Thyroxine Levothyroxine T4 and Tetraiodothyronine ) A carbon spacer arm of 10 0 A links BSA to the Thyroxine moiety Each BSA molecule contains several Thyroxine units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline pH 7 4 For in vitro research use only Not for human or animal use