Learn More
Medchemexpress LLC Thyroxine sulfate | 77074-49-8 | 99.8% | 856.93 g·mol⁻1 | C15H11I4NO7S | 100 MG
SDP
Catalog No. 5000245321
Encompass_Preferred
Thyroxine sulfate (T4 sulfate) is a sulfated thyroid hormone metabolite supplied as a research-grade reference standard for analytical and metabolic studies. It is used to study thyroid hormone metabolism, sulfation pathways, and to support assay development and calibration.
  • Sulfated thyroid hormone metabolite for research use.
  • High-purity analytical standard (listed purity 99.84%).
  • Provided as solid powder or 10 mM solution in DMSO.
  • Available in multiple sizes, including a 100 mg format.
  • Suitable for metabolic studies and assay calibration.
Catalog No. 50-002-45321 Supplier Medchemexpress LLC Supplier No. HY101406100MG
Thyroxine sulfate (T4 sulfate) is a sulfated thyroid hormone metabolite supplied as a research-grade reference standard for analytical and metabolic studies. It is used to study thyroid hormone metabolism, sulfation pathways, and to support assay development and calibration.
  • Sulfated thyroid hormone metabolite for research use.
  • High-purity analytical standard (listed purity 99.84%).
  • Provided as solid powder or 10 mM solution in DMSO.
  • Available in multiple sizes, including a 100 mg format.
  • Suitable for metabolic studies and assay calibration.