Learn More
Revvity Health Sciences Inc Thyroxine, ([125I]-T4), L-[125I]-, 500µCi (18.5MBq), Specific Activity: 5700µCi (210MBq)/µg
Catalog No. 502812325
Encompass
Thyroxine, ([125I]-T4), L-[125I]-, 500µCi (18.5MBq), Specific Activity: 5700µCi (210MBq)/µg
Catalog No. 50-281-2325 Supplier Revvity Health Sciences Inc Supplier No. NEX111X500UC
Thyroxine, ([125I]-T4), L-[125I]-, 500µCi (18.5MBq), Specific Activity: 5700µCi (210MBq)/µg