Learn More
Medchemexpress LLC HY-80013 10mg Medchemexpress, THZ1 CAS:1604810-83-4 Purity:>98%
SDP
Catalog No. 502029370
Encompass_Preferred
Medchemexpress, HY-80013 10mg THZ1 CAS:1604810-83-4 THZ1 is a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM. THZ1 also inhibits the closely related kinases CDK12 and CDK13 and downregulates MYC expression[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9370 Supplier Medchemexpress LLC Supplier No. HY8001310MG
Medchemexpress, HY-80013 10mg THZ1 CAS:1604810-83-4 THZ1 is a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM. THZ1 also inhibits the closely related kinases CDK12 and CDK13 and downregulates MYC expression[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.