Learn More
Apexbio Technology LLC THZ1 1604810-83-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A88825.1
Encompass_Preferred
THZ1 (CAS 1604810-83-4) is a covalent inhibitor of cyclin-dependent kinase 7 (CDK7) targeting specifically the cysteine residue C312 outside its kinase domain It exhibits potent inhibition with an IC50 of 3 2 nM against CDK7 in kinase binding assays In cellular studies THZ1 suppresses proliferation of Jurkat and Loucy T-cell acute lymphoblastic leukemia (T-ALL) cell lines (IC50 50 nM and 0 55 nM respectively) by disrupting CDK7-mediated signaling transcriptional regulation mediated by RUNX1 and RNA polymerase II CTD phosphorylation Thus THZ1 is utilized in cancer research to investigate transcriptional dysregulation in malignancies
Catalog No. 50-000-62319
 Restricted Item